The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145932.3 | NP_666044.1 | 2 - 3 | 494 | 85 |
GenBank mRNA | AK052725.1 | - | 2 - 3 | 495 | 85 |
AK147155.1 | - | - | 314 | 85 | |
BC024441.1 | - | 2 - 3 | 302 | 85 | |
BC025912.1 | - | 2 - 3 | 305 | 85 | |
BC031178.1 | - | 2 - 3 | 245 | 85 |