Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd33
Entrez Gene ID
208258
Gene Name
ankyrin repeat domain 33
Gene Aliases
A930021G21Rik, Panky, Panky-a
Chromosome Location
Chr.15: 101115755 - 101120407 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144790.1 NP_659039.1 3 - 4 419 74
GenBank mRNA AK044543.1 - 4 - 5 604 74
AK044546.1 - 3 - 4 397 74
BC022726.1 - 3 - 4 419 74
FJ895380.1 - 4 - 5 580 74

View Full Product Details