The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289668.1 | NP_001276597.1 | 1 - 2 | 521 | 82 |
NM_172257.4 | NP_758461.1 | 1 - 2 | 521 | 82 | |
GenBank mRNA | AK081177.1 | - | 1 - 2 | 507 | 82 |
AK152371.1 | - | 1 - 2 | 454 | 82 | |
AK155875.1 | - | 1 - 2 | 425 | 82 | |
AK196459.1 | - | 1 - 2 | 53 | 82 | |
BC023957.1 | - | 1 - 2 | 525 | 82 | |
BC051101.1 | - | 1 - 2 | 249 | 82 | |
CB247121.1 | - | 1 - 2 | 530 | 82 |