The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172412.2 | NP_766000.1 | 7 - 8 | 1287 | 67 |
GenBank mRNA | AK048605.1 | - | 5 - 6 | 2053 | 67 |
AK049639.1 | - | 7 - 8 | 1288 | 67 | |
AK160959.1 | - | 7 - 8 | 1355 | 67 | |
BC056941.1 | - | 3 - 4 | 3022 | 67 | |
BC083180.1 | - | 7 - 8 | 1294 | 67 | |
BC128572.1 | - | 3 - 4 | 1332 | 67 |