The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172532.3 | NP_766120.1 | 4 - 5 | 780 | 76 |
GenBank mRNA | AK052703.1 | - | 4 - 5 | 781 | 76 |
AK144030.1 | - | 4 - 5 | 764 | 76 | |
AK159485.1 | - | 4 - 5 | 761 | 76 | |
AK166313.1 | - | 4 - 5 | 760 | 76 | |
BC138703.1 | - | 4 - 5 | 743 | 76 |