Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Leng8
Entrez Gene ID
232798
Gene Name
leukocyte receptor cluster (LRC) member 8
Gene Aliases
AW049671
Chromosome Location
Chr.7: 4137047 - 4148177 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172736.3 NP_766324.1 3 - 4 357 92
GenBank mRNA AK034310.1 - 3 - 4 362 92
AK085304.1 - 3 - 4 355 92
AK170629.1 - 3 - 4 365 92
BC041775.1 - 3 - 4 329 92
BC042658.1 - 3 - 4 345 92
BC066768.1 - 3 - 4 348 92
CF536188.1 - 3 - 4 357 92

View Full Product Details