The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175460.3 | NP_780669.1 | 5 - 6 | 458 | 61 |
GenBank mRNA | AK083532.1 | - | 5 - 6 | 562 | 61 |
AK084041.1 | - | 5 - 6 | 568 | 61 | |
AK129155.1 | - | 5 - 6 | 544 | 61 | |
BC089007.1 | - | 5 - 6 | 458 | 61 |