Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dsg3
Entrez Gene ID
13512
Gene Name
desmoglein 3
Gene Aliases
bal, balding
Chromosome Location
Chr.18: 20510304 - 20541310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030596.3 NP_085099.2 13 - 14 2106 96
GenBank mRNA AK029018.1 - - 2128 96
AK037142.1 - 13 - 14 2137 96
AK148657.1 - 13 - 14 2107 96
BC132058.1 - 13 - 14 2135 96
BC132639.1 - 13 - 14 2135 96
U86016.1 - 13 - 14 2035 96

View Full Product Details