The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153525.5 | NP_705745.3 | 7 - 7 | 3126 | 134 |
GenBank mRNA | AK090100.1 | - | 7 - 7 | 3134 | 134 |
AK165226.1 | - | 7 - 7 | 3127 | 134 |