Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jsrp1
Entrez Gene ID
71912
Gene Name
junctional sarcoplasmic reticulum protein 1
Gene Aliases
2300003C06Rik, 2310032K21Rik, JP-45, JP45
Chromosome Location
Chr.10: 80808494 - 80813593 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028001.3 NP_082277.2 1 - 2 36 114
GenBank mRNA AK009578.1 - 1 - 2 34 114

View Full Product Details