The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081347.1 | NP_001074816.1 | 2 - 3 | 126 | 74 |
NM_001252636.1 | NP_001239565.1 | 2 - 3 | 109 | 74 | |
NM_001252637.1 | NP_001239566.1 | 2 - 3 | 230 | 74 | |
NM_001252638.1 | NP_001239567.1 | 2 - 3 | 230 | 74 | |
GenBank mRNA | AK005770.1 | - | 2 - 3 | 109 | 74 |
AK034826.1 | - | 2 - 3 | 126 | 74 | |
BB611014.1 | - | 2 - 3 | 232 | 74 | |
BC099973.1 | - | 2 - 3 | 107 | 74 | |
BC137646.1 | - | 1 - 2 | 60 | 74 | |
BC137647.1 | - | 1 - 2 | 60 | 74 | |
BC144800.1 | - | 1 - 2 | 60 | 74 |