Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dsg3
Entrez Gene ID
13512
Gene Name
desmoglein 3
Gene Aliases
bal, balding
Chromosome Location
Chr.18: 20510304 - 20541310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK029018.1 - - 2717 62

View Full Product Details