Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppme1
Entrez Gene ID
72590
Gene Name
protein phosphatase methylesterase 1
Gene Aliases
1110069N17Rik, 2700017M01Rik, PME-1, Pme1
Chromosome Location
Chr.7: 100326737 - 100371896 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028292.2 NP_082568.1 9 - 10 905 83
GenBank mRNA AK028261.1 - 9 - 10 940 83
AK076992.1 - 9 - 10 1316 83
AK146051.1 - 9 - 10 887 83
AK146268.1 - 9 - 10 917 83
AK154876.1 - 9 - 10 978 83
AK188077.1 - 3 - 4 221 83
BC014867.1 - 9 - 10 905 83
BC029064.1 - 9 - 10 893 83

View Full Product Details