Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lmod2
Entrez Gene ID
93677
Gene Name
leiomodin 2 (cardiac)
Gene Aliases
C-Lmod
Chromosome Location
Chr.6: 24597771 - 24605414 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053098.2 NP_444328.1 2 - 3 1734 118
GenBank mRNA AF237628.1 - 1 - 2 971 118
AK147141.1 - 2 - 3 1735 118

View Full Product Details