The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007577.2 | NP_001007578.2 | 5 - 5 | 1426 | 69 |
GenBank mRNA | AK140929.1 | - | 5 - 5 | 1416 | 69 |
BC067008.1 | - | 5 - 5 | 1425 | 69 |