Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fxyd6
Entrez Gene ID
59095
Gene Name
FXYD domain-containing ion transport regulator 6
Gene Aliases
0610030I18Rik, Php
Chromosome Location
Chr.9: 45370185 - 45396159 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AA521976.1 - 4 - 5 221 64
BC042579.1 - 4 - 5 228 64

View Full Product Details