Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ipo13
Entrez Gene ID
230673
Gene Name
importin 13
Gene Aliases
Imp13, Kap13, Ranbp13
Chromosome Location
Chr.4: 117894486 - 117915176 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146152.3 NP_666264.1 1 - 2 718 78
GenBank mRNA AK052257.1 - 1 - 2 719 78
AK129200.1 - 1 - 2 701 78
AK149978.1 - 1 - 2 701 78
BC031823.1 - 1 - 2 117 78

View Full Product Details