Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ipo13
Entrez Gene ID
230673
Gene Name
importin 13
Gene Aliases
Imp13, Kap13, Ranbp13
Chromosome Location
Chr.4: 117894486 - 117915176 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146152.3 NP_666264.1 18 - 19 3246 80
GenBank mRNA AK052257.1 - 18 - 19 3247 80
AK149978.1 - 18 - 19 3229 80
BC031823.1 - 18 - 19 2645 80

View Full Product Details