Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phactr4
Entrez Gene ID
100169
Gene Name
phosphatase and actin regulator 4
Gene Aliases
3110001B12Rik, AI527228, AW495572, C330013F19Rik, N28169, mKIAA4120
Chromosome Location
Chr.4: 132355922 - 132422446 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161797.1 NP_001155269.1 12 - 13 2134 66
NM_175306.4 NP_780515.2 13 - 14 2215 66
GenBank mRNA AK049209.1 - 13 - 14 2178 66
AK220496.1 - 12 - 13 2113 66
BC037059.1 - 3 - 4 301 66
BC075672.1 - 13 - 14 2177 66
BC096033.1 - 13 - 14 2134 66

View Full Product Details