Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lmtk2
Entrez Gene ID
231876
Gene Name
lemur tyrosine kinase 2
Gene Aliases
2900041G10Rik, A330101P12Rik, BREK, KPI-2, KPI2, aatyk2, cprk
Chromosome Location
Chr.5: 144100366 - 144188204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081109.1 NP_001074578.1 10 - 11 1384 82
GenBank mRNA AB288872.1 - 10 - 11 1212 82
AK158724.1 - 10 - 11 1385 82

View Full Product Details