The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163348.1 | NP_001156820.1 | 4 - 5 | 2155 | 89 |
NM_030699.2 | NP_109624.1 | 4 - 5 | 2155 | 257 | |
GenBank mRNA | AB038666.1 | - | 3 - 4 | 1060 | 89 |
AB038667.1 | - | 4 - 5 | 1932 | 257 | |
AF475069.1 | - | 3 - 4 | 1060 | 257 | |
AF475070.1 | - | 3 - 4 | 1060 | 89 |