Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpc2
Entrez Gene ID
71951
Gene Name
glypican 2 (cerebroglycan)
Gene Aliases
2410016G05Rik
Chromosome Location
Chr.5: 138273657 - 138279958 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK010515.1 - 9 - 10 1610 101

View Full Product Details