Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trmt44
Entrez Gene ID
78890
Gene Name
tRNA methyltransferase 44
Gene Aliases
2310079F23Rik, Mettl19
Chromosome Location
Chr.5: 35556208 - 35575070 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030208.3 NP_084484.2 2 - 3 626 73
GenBank mRNA AK019101.1 - 2 - 3 617 73
AK046957.1 - 2 - 3 628 73
AK132402.1 - 2 - 3 627 73
BC152332.1 - 2 - 3 621 73

View Full Product Details