Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trmt44
Entrez Gene ID
78890
Gene Name
tRNA methyltransferase 44
Gene Aliases
2310079F23Rik, Mettl19
Chromosome Location
Chr.5: 35556208 - 35575070 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030208.3 NP_084484.2 4 - 5 911 67
GenBank mRNA AK019101.1 - 4 - 5 902 67
AK046957.1 - 4 - 5 913 67
AK132402.1 - 4 - 5 912 67
BC152332.1 - 4 - 5 906 67

View Full Product Details