The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177886.1 | NP_001171357.1 | 5 - 6 | 914 | 128 |
NM_001177887.1 | NP_001171358.1 | 5 - 6 | 918 | 128 | |
NM_028078.3 | NP_082354.1 | 4 - 5 | 611 | 128 | |
GenBank mRNA | AF537215.1 | - | 5 - 6 | 852 | 128 |
AK008060.1 | - | 4 - 5 | 582 | 128 | |
AK078934.1 | - | 4 - 5 | 582 | 128 | |
AK086973.1 | - | 5 - 6 | 916 | 128 | |
BC004806.1 | - | 4 - 5 | 564 | 128 |