The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033124.1 | NP_001028296.1 | 2 - 3 | 304 | 80 |
GenBank mRNA | AK081588.1 | - | 2 - 3 | 305 | 80 |