Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdhr4
Entrez Gene ID
69398
Gene Name
cadherin-related family member 4
Gene Aliases
Cdh29
Chromosome Location
Chr.9: 107992492 - 107999683 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AI451251.1 - 1 - 2 86

View Full Product Details