Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sema3e
Entrez Gene ID
20349
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E
Gene Aliases
6430702L12, AA408817, Semah, mKIAA0331
Chromosome Location
Chr.5: 14025276 - 14256689 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011348.2 NP_035478.2 16 - 17 2514 70
GenBank mRNA AF034744.1 - 16 - 17 2249 70
AK129116.1 - 11 - 12 1279 70
AK141295.1 - 15 - 16 3237 70
AK142340.1 - 16 - 17 2341 70
AK159214.1 - 16 - 17 2305 70
BC057956.1 - 16 - 17 2412 70
Z80941.1 - 16 - 17 1880 70
Z93947.1 - 16 - 17 2492 70
Z93948.1 - 16 - 17 2492 70

View Full Product Details