Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbx22
Entrez Gene ID
245572
Gene Name
T-box 22
Gene Aliases
D230020M15Rik
Chromosome Location
Chr.X: 107667964 - 107688980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145224.3 NP_660259.1 1 - 2 42 58
GenBank mRNA AK084312.1 - 1 - 2 42 58

View Full Product Details