Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbx22
Entrez Gene ID
245572
Gene Name
T-box 22
Gene Aliases
D230020M15Rik
Chromosome Location
Chr.X: 107667964 - 107688980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145224.3 NP_660259.1 3 - 4 387 74
NM_181319.5 NP_851836.2 3 - 4 491 74
GenBank mRNA AF516208.1 - 3 - 4 491 74
AK084312.1 - 3 - 4 387 74
AY125891.1 - 2 - 3 343 74

View Full Product Details