Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbx22
Entrez Gene ID
245572
Gene Name
T-box 22
Gene Aliases
D230020M15Rik
Chromosome Location
Chr.X: 107667964 - 107688980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145224.3 NP_660259.1 4 - 5 494 66
NM_181319.5 NP_851836.2 4 - 5 598 66
GenBank mRNA AF516208.1 - 4 - 5 598 66
AK084312.1 - 4 - 5 494 66
AY125891.1 - 3 - 4 450 66

View Full Product Details