Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eral1
Entrez Gene ID
57837
Gene Name
Era (G-protein)-like 1 (E. coli)
Gene Aliases
2610524P08Rik, 9130407C09Rik, AU019798, Era, M-ERA, MERA-S, MERA-W
Chromosome Location
Chr.11: 78073376 - 78080383 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022313.2 NP_071708.2 8 - 9 1128 111
GenBank mRNA AB049389.1 - 8 - 9 1104 111
AF190730.1 - 8 - 9 1096 111
AF190731.1 - 7 - 8 864 111
AK012155.1 - 8 - 9 1118 111
BC019728.1 - 2 - 3 316 111
BC068271.1 - 8 - 9 1123 111

View Full Product Details