Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dffa
Entrez Gene ID
13347
Gene Name
DNA fragmentation factor, alpha subunit
Gene Aliases
A330085O09Rik, DFF35, Dff45, ICAD, ICAD-L, ICAD-S
Chromosome Location
Chr.4: 149104142 - 149120653 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025296.2 NP_001020467.1 1 - 2 250 74
NM_010044.3 NP_034174.2 1 - 2 250 74
NR_104263.1 1 - 2 250 74
GenBank mRNA AB009375.1 - 1 - 2 141 74
AB009376.1 - 1 - 2 141 74
AI116789.1 - 1 - 2 240 74
AK039227.1 - 1 - 2 247 74
AK051011.1 - 1 - 2 180 74
AK079634.1 - 1 - 2 208 74
AK154488.1 - 1 - 2 215 74
BC058213.1 - 1 - 2 195 74
BI736905.1 - 1 - 2 194 74
BY170336.1 - 1 - 2 251 74
BY208552.1 - 1 - 2 226 74
BY244454.1 - 1 - 2 239 74

View Full Product Details