Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Noc2l
Entrez Gene ID
57741
Gene Name
nucleolar complex associated 2 homolog (S. cerevisiae)
Gene Aliases
AA410003, AF155546, NIR
Chromosome Location
Chr.4: 156235823 - 156247616 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021303.2 NP_067278.2 2 - 3 236 107
GenBank mRNA AK076287.1 - 2 - 3 196 107
AK133834.1 - 2 - 3 236 107
AK157765.1 - 2 - 3 228 107

View Full Product Details