Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh2b1
Entrez Gene ID
20399
Gene Name
SH2B adaptor protein 1
Gene Aliases
AI425885, C530001K22Rik, Irip, Psm, SH2-B, SH2-Bb, Sh2bpsm1, mKIAA1299
Chromosome Location
Chr.7: 126466993 - 126475684 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_110346.1 4 - 5 1394 77
GenBank mRNA AA111114.1 - 3 - 4 314 77
BC011422.1 - 4 - 5 1368 77

View Full Product Details