Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgtp1
Entrez Gene ID
21822
Gene Name
T cell specific GTPase 1
Gene Aliases
Gtp2, Irgb6, Mg21, Tgtp
Chromosome Location
Chr.11: 48985329 - 48992246 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011579.3 NP_035709.3 1 - 2 241 64
GenBank mRNA AK089836.1 - 1 - 2 243 64
AK163978.1 - 1 - 2 242 64
AK172473.1 - 1 - 2 241 64
BC034256.1 - 1 - 2 64
BC085259.1 - - 212 64
BC093522.1 - 1 - 2 64
BY238768.1 - 1 - 2 244 64
U15636.1 - 1 - 2 85 64

Gene Symbol
Tgtp2
Entrez Gene ID
100039796
Gene Name
T cell specific GTPase 2
Gene Aliases
OTTMUSG00000005537
Chromosome Location
Chr.11: 49057196 - 49064212 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145164.1 NP_001138636.1 1 - 2 248 64
GenBank mRNA AK172580.1 - 1 - 2 241 64
L38444.1 - 1 - 2 79 64

View Full Product Details