Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bex2
Entrez Gene ID
12069
Gene Name
brain expressed X-linked 2
Gene Aliases
AL024066, Bex1
Chromosome Location
Chr.X: 136066565 - 136068236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009749.2 NP_033879.1 3 - 3 694 131
GenBank mRNA AK132005.1 - 3 - 3 581 131
BC027529.1 - 3 - 3 568 131

View Full Product Details