Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Srek1
- Entrez Gene ID
-
218543
- Gene Name
- splicing regulatory glutamine/lysine-rich protein 1
- Gene Aliases
-
8430401B01, AI450757, AI462342, SRrp508, SRrp86, Sfrs12, Srsf12
- Chromosome Location
-
Chr.13: 103739348 - 103774603 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK039311.1 |
- |
1 - 2 |
180 |
127 |
AK052265.1 |
- |
1 - 2 |
314 |
127 |
View Full Product Details