Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdc6
Entrez Gene ID
23834
Gene Name
cell division cycle 6
Gene Aliases
CDC18L
Chromosome Location
Chr.11: 98907702 - 98923942 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025779.1 NP_001020950.1 10 - 11 1570 82
NM_011799.2 NP_035929.1 10 - 11 1646 82
GenBank mRNA AJ009559.1 - 10 - 11 1663 82
AJ223087.1 - 10 - 11 1648 82
AK012192.1 - 10 - 11 1600 82
AK075975.1 - 10 - 11 1597 82
AK085020.1 - 10 - 11 1600 82
AK162669.1 - 11 - 12 1843 82
AK166003.1 - 10 - 11 1651 82
BC052434.1 - 10 - 11 1570 82

View Full Product Details