Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Isca2
Entrez Gene ID
74316
Gene Name
iron-sulfur cluster assembly 2 homolog (S. cerevisiae)
Gene Aliases
0710001C05Rik, 5730594E03Rik, Hbld1
Chromosome Location
Chr.12: 84773228 - 84775089 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028863.1 NP_083139.1 1 - 2 102 64
GenBank mRNA AK002936.1 - 1 - 2 101 64
BY125913.1 - 1 - 2 95 64

View Full Product Details