Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Utp6
Entrez Gene ID
216987
Gene Name
UTP6, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
4732497O03Rik, HCA66
Chromosome Location
Chr.11: 79932320 - 79962426 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144826.3 NP_659075.1 16 - 17 1584 112
GenBank mRNA AF334387.1 - 16 - 17 1911 112
AK029151.1 - 16 - 17 1585 112
AK084976.1 - 16 - 17 1577 112
AK161178.1 - 5 - 6 778 112
AK164010.1 - 16 - 17 1590 112
AK169785.1 - 16 - 17 1591 112
BC018250.1 - 16 - 17 1548 112
BC023920.1 - 16 - 17 1578 112
BC024850.1 - 16 - 17 1579 112
BC025074.1 - 4 - 5 461 112

View Full Product Details