Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
H2-Q4
Entrez Gene ID
15015
Gene Name
histocompatibility 2, Q region locus 4
Gene Aliases
H-2Q4, H2-Gs10, Qa-4, Qa4, Qat-4, Qb-1, Qb1
Chromosome Location
Chr.17: 35379555 - 35384674 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143689.1 NP_001137161.1 5 - 6 1118 67
GenBank mRNA AK037574.1 - 5 - 6 1119 67
AK136497.1 - - 1306 67
BC010602.1 - 5 - 6 1038 67

View Full Product Details