Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mlh3
Entrez Gene ID
217716
Gene Name
mutL homolog 3 (E coli)
Gene Aliases
AV125803, BB126472
Chromosome Location
Chr.12: 85234466 - 85270648 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK029786.1 - 1 - 2 1846 67

View Full Product Details