Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mab21l3
Entrez Gene ID
242125
Gene Name
mab-21-like 3 (C. elegans)
Gene Aliases
-
Chromosome Location
Chr.3: 101813076 - 101836286 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172295.4 NP_758499.3 1 - 2 305 91
GenBank mRNA AK029040.1 - 1 - 2 44 91
AK161057.1 - 1 - 2 222 91
AK172389.1 - 1 - 2 254 91
BC037703.1 - 1 - 2 242 91
CN663841.1 - 1 - 2 305 91

View Full Product Details