The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009705.3 | NP_033835.1 | 7 - 8 | 941 | 129 |
GenBank mRNA | AF032466.1 | - | 7 - 8 | 893 | 129 |
BC023349.1 | - | 7 - 8 | 905 | 129 | |
U90886.1 | - | 7 - 8 | 913 | 129 |