Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rars
Entrez Gene ID
104458
Gene Name
arginyl-tRNA synthetase
Gene Aliases
2610011N19Rik, 2610037E21Rik, AL033339, AW985894
Chromosome Location
Chr.11: 35808381 - 35834528 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025936.3 NP_080212.2 4 - 5 519 87
GenBank mRNA AK011383.1 - 4 - 5 497 87
AK076160.1 - 4 - 5 494 87
AK149856.1 - 4 - 5 494 87
AK151966.1 - 3 - 4 317 87
AK152108.1 - 4 - 5 494 87
AK160216.1 - 4 - 5 494 87
AK166097.1 - 4 - 5 494 87
AK168194.1 - 4 - 5 494 87
BC020132.1 - 4 - 5 477 87
CF748175.1 - 4 - 5 520 87

View Full Product Details