The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198724.2 | NP_942017.2 | 1 - 2 | 192 | 91 |
NM_198725.2 | NP_942018.2 | 1 - 2 | 192 | 91 | |
GenBank mRNA | AK002601.1 | - | 1 - 2 | 188 | 91 |
AY239290.1 | - | 1 - 2 | 176 | 91 | |
BC024610.1 | - | 1 - 2 | 176 | 91 | |
BY274501.1 | - | 1 - 2 | 192 | 91 |