Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alkbh3
Entrez Gene ID
69113
Gene Name
alkB, alkylation repair homolog 3 (E. coli)
Gene Aliases
1700108H04Rik, 1810020C19Rik, Abh3, mABH3
Chromosome Location
Chr.2: 93980634 - 94010823 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026944.1 NP_081220.1 7 - 8 578 100
GenBank mRNA BC018196.1 - 7 - 8 578 100

View Full Product Details