The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027163.4 | NP_081439.1 | 2 - 3 | 158 | 73 |
GenBank mRNA | AK009787.1 | - | 2 - 3 | 170 | 73 |
AY071842.1 | - | 2 - 3 | 143 | 73 | |
BC119517.1 | - | 2 - 3 | 158 | 73 |