Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Top3b
Entrez Gene ID
21976
Gene Name
topoisomerase (DNA) III beta
Gene Aliases
-
Chromosome Location
Chr.16: 16870694 - 16892986 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011624.2 NP_035754.1 7 - 8 1020 79
GenBank mRNA AB013603.1 - 6 - 7 833 79
AB045324.1 - 7 - 8 1094 79
AK041159.1 - 7 - 8 930 79
AK144709.1 - 8 - 9 1006 79
AK146475.1 - 7 - 8 1042 79
BC031723.1 - 7 - 8 1020 79

View Full Product Details